missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP2R1A Recombinant Rabbit Monoclonal Antibody (HL1781)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA547179

Product Code. 17849695

  • € 474.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The PR65 subunit of protein phosphatase 2A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. Upon interaction with GNA12 promotes dephosphorylation of microtubule associated protein TAU/MAPT. Required for proper chromosome segregation and for centromeric localization of SGO1 in mitosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PPP2R1A
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P30153, Q76MZ3
PPP2R1A
Recombinant fragment of human PPP2R1A.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot
HL1781
Unconjugated
PPP2R1A
29B; 6330556D22Rik; 65 kDa regulatory subunit of protein phosphatase; A/PR65; alpha isoform of regulatory subunit A, protein phosphatase 2; CG13383; CG17291; CG17291-PA; CG17291-PB; CG17291-PC; CG17291-PD; CG17291-PE; CG33297; Dmel\CG17291; Dmel_CG17291; dPP2A A; DPR65; Medium tumor antigen-associated 61 kDa protein; MGC786; MRD36; PP2A; PP2A 29B; PP2A 65kD subunit A; PP2A A; PP2A subunit A isoform PR65-alpha; PP2A subunit A isoform R1-alpha; PP2A[A]; Pp2A29B; PP2A-29B; Pp2A-29B-PA; Pp2A-29B-PB; Pp2A-29B-PC; Pp2A-29B-PD; Pp2A-29B-PE; PP2A-A; PP2AAALPHA; PP2A-Aalpha; PP2Ac; PP2CA; PP2Calpha; Ppp2r1a; PR65; PR65A; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A , alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A, alpha isoform; protein phosphatase 2 regulatory subunit A, alpha; protein phosphatase 2 scaffold subunit Aalpha; protein phosphatase 2, regulatory subunit A, alpha; protein phosphatase 2A; protein phosphatase 2A at 29B; protein phosphatase PP2A; Protein phosphatase PP2A regulatory subunit A; protein phosphate 2A; regulatory subunit A of type 2A protein phosphatase; RP-C; serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alpha isoform; serine/threonine protein phosphatase A subunit type 2A; serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alpha isoform; Serine/threonine-protein phosphatase PP2A 65 kDa regulatory subunit; testicular secretory protein Li 1; Unknown (protein for MGC:128399)
Rabbit
Protein A
RUO
117281, 51792, 5518
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.