missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R1B Monoclonal Antibody (OTI8C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811459
This item is not returnable.
View return policy
Description
In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase (PP) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit and a catalytic subunit. Four major families of protein phosphatase catalytic subunits have been identified, designated PP1, PP2A, PP2B (calcineurin) and PP2C. An additional protein phosphatase catalytic subunit, PPX (also known as PP4), is a putative member of a novel PP family. The PP2A family comprises subfamily members PP2A alpha and PP2A beta. The PP2A catalytic subunit associates with a variety of regulatory subunits. Regulatory subunits include PP2A-A alpha and -A beta, PP2A-B alpha and -B beta, PP2A-C alpha and -C beta, and PP2A-B56 alpha and -B56 beta.Specifications
| PPP2R1B | |
| Monoclonal | |
| Unconjugated | |
| PPP2R1B | |
| PP2A-Abeta, PR65B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5519 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI8C5 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P30154 | |
| PPP2R1B | |
| Human recombit protein fragment corresponding to amino acids 19-601 of human PPP2R1B produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction