missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP2R3A Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142323
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognise both reported isoforms (NP_002709.2; NP_871626.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Protein phosphatase 2 (formerly named type 2A) is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B'/PR61, and B'/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holoenzyme. The product of this gene belongs to the B' family. The B' family has been further divided into subfamilies. The product of this gene belongs to the alpha subfamily of regulatory subunit B'. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| PPP2R3A | |
| Polyclonal | |
| Unconjugated | |
| PPP2R3A | |
| 3222402P14Rik; A730042E07; DNA for thyroid hormone receptor binding site (258bp); PP2A subunit B isoform PR72/PR130; PP2A subunit B isoform R3 isoform; PP2A subunit B isoforms B72/B130; PP2A subunit B isoforms B''-PR72/PR130; PP2A, subunit B, R3 isoform; PPP2R3; PPP2R3A; PR130; PR72; protein phosphatase 2 (formerly 2A), regulatory subunit B'' (PR 72), alpha isoform and (PR 130), beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B'', alpha; protein phosphatase 2 regulatory subunit B'', alpha; protein phosphatase 2 regulatory subunit B''alpha; protein phosphatase 2, regulatory subunit B'', alpha; serine/threonine-protein phosphatase 2A 72/130 kDa regulatory subunit B; Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit alpha; Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit alpha-like protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 5523 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q06190 | |
| PPP2R3A | |
| Peptide with sequence C-QKDVENDGPEPSD. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction