missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP2R3B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA514873

Product Code. 10868860

  • € 520.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PPP2R3B Polyclonal Antibody for Western Blot

Protein phosphatase 2 is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B'/PR61, and B'/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B' family. The B' family has been further divided into subfamilies. The product of this gene belongs to the beta subfamily of regulatory subunit B'. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PPP2R3B
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q9Y5P8
PPP2R3B
KLH conjugated synthetic peptide between 25-54 amino acids from the N-terminal region of human PR48.
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
PPP2R3B
NYREN8; NY-REN-8; NY-REN-8 antigen; PP2A subunit B isoform PR48; PP2A, subunit B, PR48 isoform; PPP2R3B; PPP2R3L; PPP2R3LY; PR48; protein phosphatase 2 (formerly 2A), regulatory subunit B', beta; protein phosphatase 2 regulatory subunit B', beta; protein phosphatase 2 regulatory subunit B'beta; protein phosphatase 2, regulatory subunit B', beta; Protein phosphatase 2A 48 kDa regulatory subunit; serine/threonine protein phosphatase 2A, 48kDa regulatory subunit B; serine/threonine-protein phosphatase 2A regulatory subunit B' subunit beta
Rabbit
Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
RUO
28227
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.