missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP2R5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140022
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: TLNELVEYVS TNRGVIVESA YSD Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an alpha isoform of the regulatory subunit B56 subfamily. Alternative transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| PPP2R5A | |
| Polyclonal | |
| Unconjugated | |
| PPP2R5A | |
| B56A; MGC131915; PP2A B subunit isoform B56-alpha; PP2A B subunit isoform B'-alpha; PP2A B subunit isoform PR61-alpha; PP2A B subunit isoform R5-alpha; PP2A, B subunit, B' alpha isoform; PP2A, B subunit, B56 alpha isoform; PP2A, B subunit, PR61 alpha isoform; PP2A, B subunit, R5 alpha isoform; PP2CB; PPP2R5A; PR61A; PR61alpha; protein phosphatase 2 regulatory subunit B', alpha; protein phosphatase 2 regulatory subunit B'alpha; protein phosphatase 2, regulatory subunit B (B56), alpha isoform; protein phosphatase 2, regulatory subunit B', alpha; protein phosphatase 2, regulatory subunit B', alpha isoform; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, alpha isoform; serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit alpha isoform | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5525 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15172 | |
| PPP2R5A | |
| Recombinant protein fragment of Human PPP2R5A (Product #RP-106923) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction