missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R5B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93189-0.1ml
This item is not returnable.
View return policy
Description
PPP2R5B Polyclonal antibody specifically detects PPP2R5B in Human samples. It is validated for Western Blot
Specifications
| PPP2R5B | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| B56B, FLJ35411, PP2A B subunit isoform B56-beta, PP2A B subunit isoform B'-beta, PP2A B subunit isoform PR61-beta, PP2A B subunit isoform R5-beta, PR61B, protein phosphatase 2, regulatory subunit B (B56), beta isoform, protein phosphatase 2, regulatory subunit B', beta, protein phosphatase 2, regulatory subunit B', beta isoform, serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, betaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit beta isoform | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human PPP2R5B (NP_006235.1). METKLPPASTPTSPSSPGLSPVPPPDKVDGFSRRSLRRARPRRSHSSSQFRYQSNQQELTPLPLLKDVPASELHELLSRKLAQCGVMFDFLDCVADLKGKEVKRAALNELVECVGSTRGVLIEPVYPDII | |
| 0.1 mL | |
| Wnt Signaling Pathway | |
| 5526 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction