missing translation for 'onlineSavingsMsg'
Learn More

PPP2R5C Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-94154-0.1ml

 View more versions of this product

Product Code. 18638741

  • € 470.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PPP2R5C Polyclonal antibody specifically detects PPP2R5C in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PPP2R5C
Polyclonal
Western Blot 1:500 - 1:2000
B' alpha regulatory subunit, B56G, KIAA0044, MGC23064, PP2A B subunit isoform B56-gamma, PP2A B subunit isoform B'-gamma, PP2A B subunit isoform PR61-gamma, PP2A B subunit isoform R5-gamma, PR61G, protein phosphatase 2, regulatory subunit B (B56), gamma isoform, protein phosphatase 2, regulatory subunit B', gamma, protein phosphatase 2, regulatory subunit B', gamma isoform, Renal carcinoma antigen NY-REN-29, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit gamma isoform
Recombinant fusion protein containing a sequence corresponding to amino acids 1-320 of human PPP2R5C (NP_848702.1). MLTCNKAGSRMVVDAANSNGPFQPVVLLHIRDVPPADQEKLFIQKLRQCCVLFDFVSDPLSDLKWKEVKRAALSEMVEYITHNRNVITEPIYPEVVHMFAVNMFRTLPPSSNPTGAEFDPEEDEPTLEAAWPHLQLVYEFFLRFLESPDFQPNIAKKYIDQKFVLQLLELFDSEDPRERDFLKTTLHRIYGKFLGLRAYIRKQINNIFYRFIYETEHHNGIAELLEILGSIINGFALPLKEEHKIFLLKVLLPLHKVKSLSVYHPQLAYCVVQFLEKDSTLTEPVVMALLKYWPKTHSPKEVMFLNELEEILDVIEPSEF
0.1 mL
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Phosphatase, Signal Transduction, Wnt Signaling Pathway
5527
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.