Learn More
Novus Biologicals™ PPP2R5D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30776-100ug
 View more versions of this product
Product Code. 30067127
  • € 508.00 / 100µg
Description
PPP2R5D Polyclonal antibody specifically detects PPP2R5D in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PPP2R5D
Polyclonal
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
Unconjugated
MGC2134, MGC8949, PP2A B subunit isoform B56-delta, PP2A B subunit isoform B'-delta, PP2A B subunit isoform PR61-delta, PP2A B subunit isoform R5-delta, PP2A, B subunit, B' delta isoform, PP2A, B subunit, B56 delta isoform, PP2A, B subunit, PR61 delta isoform, PP2A, B subunit, R5 delta isoform, protein phosphatase 2, regulatory subunit B', delta, regulatory subunit B (B56), delta isoform, regulatory subunit B', delta isoform, Serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, deltaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform
Synthetic peptide conjugated to KLH derived from the N-terminus of PPP2R5D
100 μg
Neuroscience, Wnt Signaling Pathway
5528
Store at -20°C. Avoid freeze/thaw cycles.