missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R5D Antibody (OTI5E7), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73587AF594
This item is not returnable.
View return policy
Description
PPP2R5D Monoclonal specifically detects PPP2R5D in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PPP2R5D | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PPP2R5D | |
| Full length recombinant protein of human PPP2R5D (NP_006236) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI5E7 | |
| Western Blot, Immunohistochemistry | |
| MGC2134, MGC8949, PP2A B subunit isoform B56-delta, PP2A B subunit isoform B′-delta, PP2A B subunit isoform PR61-delta, PP2A B subunit isoform R5-delta, PP2A, B subunit, B′ delta isoform, PP2A, B subunit, B56 delta isoform, PP2A, B subunit, PR61 delta isoform, PP2A, B subunit, R5 delta isoform, protein phosphatase 2, regulatory subunit B′, delta, regulatory subunit B (B56), delta isoform, regulatory subunit B′, delta isoform, Serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, deltaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform | |
| Mouse | |
| Affinity Purified | |
| Neuroscience, Wnt Signaling Pathway | |
| 5528.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction