missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R5E Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 468.00
Specifications
| Antigen | PPP2R5E |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PPP2R5E Polyclonal antibody specifically detects PPP2R5E in Human, Mouse samples. It is validated for Western BlotSpecifications
| PPP2R5E | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Wnt Signaling Pathway | |
| PBS (pH 7.3), 50% glycerol | |
| 5529 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| epsilon isoform of regulatory subunit B56, protein phosphatase 2A, PP2A B subunit isoform B56-epsilon, PP2A B subunit isoform B'-epsilon, PP2A B subunit isoform PR61-epsilon, PP2A B subunit isoform R5-epsilon, PP2A, B subunit, B' epsilon, PP2A, B subunit, B56 epsilon, PP2A, B subunit, PR61 epsilon, PP2A, B subunit, R5 epsilon, protein phosphatase 2, regulatory subunit B (B56), epsilon isoform, protein phosphatase 2, regulatory subunit B', epsilon isoform, serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, epsilon, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit epsilonisoform | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human PPP2R5E (NP_006237.1). LLKDVPSSEQPELFLKKLQQCCVIFDFMDTLSDLKMKEYKRSTLNELVDYITISRGCLTEQTYPEVVRMVSCNIFRTLPPSDSNEFDPEEDEPTLEASWPH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title