Learn More
PPP3R2 (Human) Matched Antibody Pair, Abnova™
Matched Antibody Pair
Brand:  Abnova H00005535-AP11.1Set
Product Code. 16142454
  • € 843.00 / 1 set
Specifications
PPP3R2
Unconjugated
PPP3RL
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PPP3R2 (100 ug) 2. Detection antibody: mouse monoclonal anti-PPP3R2, IgG2a Kappa (20 ug) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
ELISA
protein phosphatase 3 (formerly 2B), regulatory subunit B, beta isoform
PPP3R2
RUO
5535
Human
Ab Pair