missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP4C Monoclonal Antibody (OTI2H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810212S
This item is not returnable.
View return policy
Description
PPP4C Monoclonal antibody specifically detects PPP4C in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PPP4C | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96PU4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 115426 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2H8 | |
| Unconjugated | |
| UHRF2 | |
| 1110002D08Rik; AU016079; P PPX; pp4; PP4C; PPH3; Ppp4; PPP4C; PPX; PP-X; protein phosphatase 4 (formerly X), catalytic subunit; protein phosphatase 4 catalytic subunit; protein phosphatase 4, catalytic subunit; protein phosphatase X; protein phosphatase X, catalytic subunit; serine/threonine-protein phosphatase 4 catalytic subunit | |
| Human recombinant protein fragment corresponding to amino acids 1-288 of human UHRF2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction