missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP4R1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104131
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PPP4R1.
Regulatory subunit of serine/threonine-protein phosphatase 4. May play a role in regulation of cell division in renal glomeruli. The PPP4C-PPP4R1 PP4 complex may play a role in dephosphorylation and regulation of HDAC3.
Specifications
| PPP4R1 | |
| Polyclonal | |
| Unconjugated | |
| PPP4R1 | |
| 3110001J10Rik; MEG1; meg-1; PP4(Rmeg); PP4R1; Ppp4r1; protein phosphatase 4 regulatory subunit 1; protein phosphatase 4, regulatory subunit 1; serine/threonine phosphatase 4; serine/threonine-protein phosphatase 4 regulatory subunit 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140943, 70351, 9989 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8K2V1, Q8TF05, Q8VI02 | |
| PPP4R1 | |
| A synthesized peptide derived from human PPP4R1(Accession Q8TF05), corresponding to amino acid residues I164-L214. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction