missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP4R2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19915
This item is not returnable.
View return policy
Description
PPP4R2 Polyclonal specifically detects PPP4R2 in Human samples. It is validated for Western Blot.
Specifications
| PPP4R2 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| PPP4R2 | |
| Recombinant protein encompassing a sequence within the center region of human PPP4R2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 151987 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:1000-1:10000 | |
| MGC131930, PP4R2, protein phosphatase 4, regulatory subunit 2, serine/threonine-protein phosphatase 4 regulatory subunit 2 | |
| Rabbit | |
| 40 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction