missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP4R4 Monoclonal Antibody (OTI1G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810585S
This item is not returnable.
View return policy
Description
PPP4R4 Monoclonal antibody specifically detects PPP4R4 in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| PPP4R4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P63329, Q08209 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 24674, 5530 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI1G7 | |
| Unconjugated | |
| PPP3CA | |
| 8430415E04Rik; CFAP14; cilia and flagella associated protein 14; HEAT-like repeat-containing protein; KIAA1622; mKIAA1622; PP4R4; PPP4R4; protein phosphatase 4 regulatory subunit 4; protein phosphatase 4, regulatory subunit 4; RGD1560932; Serine/threonine-protein phosphatase 4 regulatory subunit 4 | |
| Human recombinant protein fragment corresponding to amino acids 2-105 of human PPP3CA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction