missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP5C Mouse anti-Canine, Human, Clone: OTI4G8, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500592
This item is not returnable.
View return policy
Description
This gene encodes a serine/threonine phosphatase which is a member of the protein phosphatase catalytic subunit family. Proteins in this family participate in pathways regulated by reversible phosphorylation at serine and threonine residues; many of these pathways are involved in the regulation of cell growth and differentiation. The product of this gene has been shown to participate in signaling pathways in response to hormones or cellular stress, and elevated levels of this protein may be associated with breast cancer development. Alternative splicing results in multiple transcript variants.Specifications
| PPP5C | |
| Monoclonal | |
| 0.9 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P53041 | |
| PPP5C | |
| Full length human recombit protein of human PPP5C produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4G8 | |
| Unconjugated | |
| PPP5C | |
| PP5, PPP5, PPT | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5536, 612199 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction