missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP6C Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2883AF750
This item is not returnable.
View return policy
Description
PPP6C Polyclonal antibody specifically detects PPP6C in Human, Mouse samples. It is validated for Western Blot
Specifications
| PPP6C | |
| Polyclonal | |
| Western Blot | |
| EC 3.1.3.16, FLJ92648, MGC12249, PP6, PP6C, PPP6, protein phosphatase 6, catalytic subunit, serine/threonine protein phosphatase catalytic subunit, serine/threonine-protein phosphatase 6 catalytic subunit | |
| The immunogen recognized by this antibody maps to a region between residue 250 and the C-terminus (residue 305) of human Protein Phosphatase 6, Catalytic Subunit using the numbering given in entry NP_002712.1 (GeneID 5537). | |
| 0.1 mL | |
| Cell Cycle and Replication, Signal Transduction | |
| 5537 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction