missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP6C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 415.00 - € 589.00
Specifications
| Antigen | PPP6C |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18406431
|
Novus Biologicals
NBP2-13804-25ul |
25ul |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18099318
|
Novus Biologicals
NBP2-13804 |
0.1 mL |
€ 589.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPP6C Polyclonal specifically detects PPP6C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PPP6C | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5537 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: CYRCGNIASIMVFKDVNTREPKLFRAVPDSERVIPPRTTTPYFL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 3.1.3.16, FLJ92648, MGC12249, PP6, PP6C, PPP6, protein phosphatase 6, catalytic subunit, serine/threonine protein phosphatase catalytic subunit, serine/threonine-protein phosphatase 6 catalytic subunit | |
| PPP6C | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title