missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPPDE1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76320AF647
This item is not returnable.
View return policy
Description
PPPDE1 Polyclonal antibody specifically detects PPPDE1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| PPPDE1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 647 | |
| C1orf121, CGI-146, chromosome 1 open reading frame 121, FAM152A, FLJ21998, member A, PNAS-4, PPPDE peptidase domain containing 1, PPPDE peptidase domain-containing protein 1, Protein FAM152A | |
| Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human PNAS4. The immunogen is located within the last 50 amino acids of PNAS4. Amino Acid Squence: VSQELQDELEEAEDAAAS | |
| 0.1 mL | |
| Cell Biology | |
| 51029 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction