missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPT1 Antibody (OTI1F10), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73588PE
This item is not returnable.
View return policy
Description
PPT1 Monoclonal antibody specifically detects PPT1 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| PPT1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1F10 | |
| Western Blot, Immunohistochemistry | |
| ceroid-palmitoyl-palmitoyl-protein thioesterase 1, CLN1EC 3.1.2.22, INCL, Palmitoyl-protein hydrolase 1, palmitoyl-protein thioesterase 1, PPT, PPT-1 | |
| Human recombinant protein fragment corresponding to amino acids 100-306 of human PPT1 (NP_000301) produced in E.coli. | |
| 0.1 mL | |
| Neuroscience | |
| 5538 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction