missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPT1 Monoclonal Antibody (OTI1F10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526471
This item is not returnable.
View return policy
Description
PPT1 Monoclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene is a small glycoprotein involved in the catabolism of lipid-modified proteins during lysosomal degradation. The encoded enzyme removes thioester-linked fatty acyl groups such as palmitate from cysteine residues. Defects in this gene are a cause of infantile neuronal ceroid lipofuscinosis 1 (CLN1, or INCL) and neuronal ceroid lipofuscinosis 4 (CLN4). Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| PPT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50897 | |
| Ppt1 | |
| Human recombinant protein fragment corresponding to amino acids 100-306 of PPT1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1F10 | |
| Unconjugated | |
| Ppt1 | |
| 3.1.2.22; 9530043G02Rik; AA960502; C77813; ceroid-palmitoyl-palmitoyl-protein thioesterase 1; CLN1; D4Ertd184e; INCL; Palmitoyl-protein hydrolase 1; palmitoyl-protein thioesterase; palmitoyl-protein thioesterase 1; palmitoyl-protein thioesterase 1 (ceroid-lipofuscinosis, neuronal 1, infantile); PPT; Ppt1; PPT-1; QnpA-18851; unnamed protein product | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5538 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction