missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPT2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94272-0.1ml
This item is not returnable.
View return policy
Description
PPT2 Polyclonal antibody specifically detects PPT2 in Mouse samples. It is validated for Western Blot
Specifications
| PPT2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| C6orf8, DKFZp564P1516, EC 3.1.2, EC 3.1.2.-, G14, lysosomal thioesterase PPT2, palmitoyl-protein hydrolase 2, palmitoyl-protein thioesterase 2, PPT-2, S-thioesterase G14, VE-statin2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 28-302 of human PPT2 (NP_005146.4). APAPHRASYKPVIVVHGLFDSSYSFRHLLEYINETHPGTVVTVLDLFDGRESLRPLWEQVQGFREAVVPIMAKAPQGVHLICYSQGGLVCRALLSVMDDHNVDSFISLSSPQMGQYGDTDYLKWLFPTSMRSNLYRICYSPWGQEFSICNYWHDPHHDDLYLNASSFLALINGERDHPNATVWRKNFLRVGHLVLIGGPDDGVITPWQSSFFGFYDANETVLEMEEQLVYLRDSFGLKTLLARGAIVRCPMAGISHTAWHSNRTLYETCIEPWLS | |
| 0.1 mL | |
| Neurodegeneration, Neuroscience | |
| 9374 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction