Learn More
Grant Instruments™ PQ Documentation
Click to view available options
For Use With (Equipment):
All BTD Models
All Grant XUB Digital Ultrasonic Baths
All Grant XUBA Analogue Ultrasonic Baths
All JB Aqua Plus Models (JBN and JBA)
All QBD Models
All SUB Aqua Pro Models (SAP)
Grant CS200G Stirred Thermostatic Bath/Circulator
Grant CVP-2 PCR Plate Centrifuge
Grant CXG Stirred Thermostatic Bath/Circulator
Grant PHMP-100 Thermoshaker for Microplates
Grant R4 Refrigerated Baths/Circulators
Grant T100 Stirred Thermostatic Bath/Circulator
Grant TC120 Stirred Thermostatic Bath/Circulator
Grant TS-DW Deep Well Plate Thermoshaker
Grant TX150 Stirred Thermostatic Bath/Circulator
Grant TXF200 Stirred Thermostatic Bath/Circulator
Unit Size:
Each
Description
Performance Qualification (PQ): Instrument is continuing to meet its specification
- Each documentation set is specific to individual serial numbered equipment and is provided in an easy to use ring bound format.
- PQ documentation can be ordered at any time.
Specifications
Specifications
| For Use With (Equipment) | All QBD Models |
| Product Type | PQ Documentation |