missing translation for 'onlineSavingsMsg'
Learn More

PR 619, TRC

Product Code. 30415656
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415656 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415656 Supplier Toronto Research Chemicals Supplier No. TRCP69901025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2,6-diamino-5-thiocyanatopyridin-3-yl) thiocyanate
CAS 2645-32-1
Molecular Formula C7 H5 N5 S2
Quantity 25 mg
Synonym Thiocyanic Acid C,C'-(2,6-Diamino-3,5-pyridinediyl) Ester, 2,6-Diamino-3,5-dithiocyanopyridine
InChI Formula InChI=1S/C7H5N5S2/c8-2-13-4-1-5(14-3-9)7(11)12-6(4)10/h1H,(H4,10,11,12)
SMILES Nc1nc(N)c(SC#N)cc1SC#N
IUPAC Name (2,6-diamino-5-thiocyanatopyridin-3-yl) thiocyanate
Molecular Weight (g/mol) 223.278
Formula Weight 222.999
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.