Learn More
Tocris Bioscience™ PR 619
GREENER_CHOICE
Broad spectrum DUB inhibitor
Brand:  Tocris Bioscience™ 4482/50
Encompass
Product Code. 15776186
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
PR 619 is a broad spectrum, reversible DUB inhibitor. Exhibits limited activity against other proteases. Induces accumulation of polyubiquitinated proteins, but has no direct inhibitory effect on the proteasome. Cytotoxic in HEK293T and colorectal cancer cells. Stabilizes microtubule network in oligodendroglial cells.
Specifications
Specifications
PR 619
2,6-Diamino-3,5-dithiocyanopyridine
Soluble to 10mM in DMSO
Bioactive Small Molecule
2645-32-1
50 mg
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Title
Select an issue
Research Use Only. Not For Human or Veterinary Use