missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAC Antibody, Alexa Fluor™ 594 , Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97053AF594
This item is not returnable.
View return policy
Description
PRAC Polyclonal antibody specifically detects PRAC in Human samples. It is validated for ELISA
Specifications
| PRAC | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Alexa Fluor 594 | |
| C17orf92, chromosome 17 open reading frame 92, MGC32520, PRAC1, prostate cancer susceptibility candidate, Prostate, rectum and colon expressed gene protein, small nuclear protein PRAC | |
| Produced in rabbits immunized with purified, recombinant Human PRAC (NP_115767.1; Met1-Phe57). | |
| 0.1 mL | |
| Cancer, Cell Biology, Tumor Suppressors | |
| 84366 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction