missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PRAF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29931
This item is not returnable.
View return policy
Description
PRAF1 Polyclonal antibody specifically detects PRAF1 in Human samples. It is validated for Immunoprecipitation
Specifications
| PRAF1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 64425 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| DNA-directed RNA polymerase I subunit E, DNA-directed RNA polymerase I subunit RPA49, FLJ13390, FLJ13970, FLJ43482, PAF53RNA polymerase I-associated factor 1, polymerase (RNA) I associated factor 1, polymerase (RNA) I polypeptide E, 53kDa, PRAF1, RNA polymerase I associated factor 53, RNA polymerase I subunit A49, RNA polymerase I-associated factor 53, RP11-405L18.3 | |
| Between 431 and 481 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction