missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prajmalium Bitartrate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Prajmalium Bitartrate |
| CAS | 2589-47-1 |
| Analyte or Component Names | Prajmalium Bitartrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H33 N2 O2 . C4 H5 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ajmalanium, 17,21-dihydroxy-4-propyl-, (17R,21α)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1), Ajmalanium, 17,21-dihydroxy-4-propyl-, (17R,21α)-, salt with (2R,3R)-2,3-dihydroxybutanedioic acid (1:1) (9CI), Ajmalanium, 17,21-dihydroxy-4-propyl-, (17R,21α)-, salt with [R-(R*,R*)]-2,3-dihydroxybutanedioic acid (1:1), 5H-6,10:11,12a-Dimethanoindolo[3,2-b]quinolizine, ajmalanium deriv., Butanedioic acid, 2,3-dihydroxy- (2R,3R)-, ion(1-), (17R,21α)-17,21-dihydroxy-4-propylajmalanium (9CI), Butanedioic acid, 2,3-dihydroxy-[R-(R*,R*)]-, ion(1-), (17R,21α)-17,21-dihydroxy-4-propylajmalanium, GT 1012, N-Propylajmaline bitartrate, N-Propylajmaline dihydrogen tartrate, N-Propylajmaline hydrogen tartrate, N-Propylajmalinium hydrogen tartrate, N-n-Propylajmalinium hydrogen tartrate, NPAB, Neo-Gilurytmal, Prajmaline bitartrate, Prajmalium bitartrate |
| InChI Formula | InChI=1S/C23H33N2O2.C4H6O6/c1-4-10-25-17-11-14(13(5-2)22(25)27)19-18(25)12-23(21(19)26)15-8-6-7-9-16(15)24(3)20(17)23;5-1(3(7)8)2(6)4(9)10/h6-9,13-14,17-22,26-27H,4-5,10-12H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/q+1;/p-1/t13-,14-,17-,18-,19+,20-,21+,22+,23+,25-;1-,2-/m01/s1 |
| Solution Type | Neat |
| SMILES | CCC[N@+]12[C@H](O)[C@@H](CC)[C@@H]3C[C@H]1[C@@H]4N(C)c5ccccc5[C@]46C[C@H]2[C@H]3[C@H]6O.O[C@H]([C@@H](O)C(=O)[O-])C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?