missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAME Antibody (CL5148), Novus Biologicals™
Mouse Monoclonal Antibody
€ 369.00 - € 500.00
Specifications
| Antigen | PRAME |
|---|---|
| Clone | CL5148 |
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:250 - 1:500, Immunohistochemistry-Paraffin 1:250 - 1:500 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18202724
|
Novus Biologicals
NBP2-61149 |
100 μL |
€ 500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645656
|
Novus Biologicals
NBP2-61149-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRAME Monoclonal specifically detects PRAME in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PRAME | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:250 - 1:500, Immunohistochemistry-Paraffin 1:250 - 1:500 | |
| Unconjugated | |
| Mouse | |
| Human | |
| PRAME | |
| IgG1 | |
| Protein A purified |
| CL5148 | |
| Monoclonal | |
| Purified | |
| Cancer | |
| 23532 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LLLSHIHASSYISPEKEEQYIAQFTSQFLSLQCLQALYVDSLFFLRGRLDQLLRHVMNPLETLSITNCRLSEGDVMHLSQSPSVSQLSVLSLSGVMLTDVSPEPLQALLERASATLQDLVFDECGI | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title