missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAME Antibody (PRAME/6928R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 311.00 - € 565.00
Specifications
| Antigen | PRAME |
|---|---|
| Clone | PRAME/6928R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687134
|
Novus Biologicals
NBP3-20237-20ug |
20 μg |
€ 311.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18634784
|
Novus Biologicals
NBP3-20237-100ug |
100 μg |
€ 565.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRAME Monoclonal antibody specifically detects PRAME in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| PRAME | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 10mM PBS with 0.05% BSA | |
| 23532 | |
| IgG κ | |
| Protein A or G purified |
| PRAME/6928R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cancer/testis antigen 130, MAPEOIP4CT130, melanoma antigen preferentially expressed in tumors, OIP-4, Opa-interacting protein 4, Opa-interacting protein OIP4, preferentially expressed antigen in melanoma, Preferentially expressed antigen of melanoma | |
| Recombinant fragment (around aa100-509) of human PRAME protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title