missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PRAME Antibody (PRAME/9188) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26932JF549
This item is not returnable.
View return policy
Description
PRAME Monoclonal antibody specifically detects PRAME in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PRAME | |
| Monoclonal | |
| Janelia Fluor 549 | |
| cancer/testis antigen 130, MAPEOIP4CT130, melanoma antigen preferentially expressed in tumors, OIP-4, Opa-interacting protein 4, Opa-interacting protein OIP4, preferentially expressed antigen in melanoma, Preferentially expressed antigen of melanoma | |
| Recombinant fragment (around aa100-250) of human PRAME protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer | |
| 23532 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| PRAME/9188 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction