missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PRAME Antibody (PRAME/9188), Novus Biologicals™

Mouse Monoclonal Antibody

€ 245.00 - € 486.00

Specifications

Antigen PRAME
Clone PRAME/9188
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043309
View Documents
Novus Biologicals™
NBP3-26783-100ug
100 μg
€ 486.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043241
View Documents
Novus Biologicals™
NBP3-26783-20ug
20 μg
€ 245.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PRAME Monoclonal antibody specifically detects PRAME in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

PRAME
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer
10 mM PBS with 0.05% BSA
23532
IgG2b κ
Protein A or G purified
PRAME/9188
Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Monoclonal
Purified
RUO
Human
cancer/testis antigen 130, MAPEOIP4CT130, melanoma antigen preferentially expressed in tumors, OIP-4, Opa-interacting protein 4, Opa-interacting protein OIP4, preferentially expressed antigen in melanoma, Preferentially expressed antigen of melanoma
Recombinant fragment (around aa100-250) of human PRAME protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.