Learn More
PRAME Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528394
Description
PA5-28394 targets PRAME in WB applications and shows reactivity with Human samples. The PA5-28394 immunogen is recombit fragment corresponding to a region within amino acids 79 and 386 of Human PRAME. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
This gene encodes an antigen that is predomitly expressed in human melanomas and that is recognized by cytolytic T lymphocytes. It is not expressed in normal tissues, except testis. This expression pattern is similar to that of other CT antigens, such as MAGE, BAGE and GAGE. However, unlike these other CT antigens, this gene is also expressed in acute leukemias. Five alternatively spliced transcript variants encoding the same protein have been observed for this gene.
Specifications
| PRAME | |
| Polyclonal | |
| Unconjugated | |
| P78395 | |
| PRAME | |
| Recombit fragment corresponding to a region within amino acids 79 and 386 of Human PRAME | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| PRAME | |
| CT130, MAPE, OIP4, OIP-4, Opa-interacting protein OIP4, cancer/testis antigen 130, melanoma antigen preferentially expressed in tumors, opa-interacting protein 4, preferentially expressed antigen of melanoma | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23532 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.