missing translation for 'onlineSavingsMsg'
Learn More

Pramoxine Hydrochloride, TRC

Product Code. 30412931
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412931 10 g 10g
30421353 1 g -
30409065 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30412931 Supplier Toronto Research Chemicals Supplier No. TRCP70085010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-58-1
Molecular Formula C17 H27 N O3 . Cl H
Molecular Weight (g/mol) 329.86
Synonym Morpholine, 4-[3-(4-butoxyphenoxy)propyl]-, hydrochloride (1:1)Morpholine, 4-[3-(4-butoxyphenoxy)propyl]-, hydrochloride (9CI), Morpholine, 4-[3-(p-butoxyphenoxy)propyl]-, hydrochloride (7CI,8CI), Pramoxine hydrochloride (6CI), 4-[3-(p-Butoxyphenoxy)propyl]morpholine hydrochloride, Pramacort, Pramocaine hydrochloride, Proctofoam, Tronolaine, Tronothan, Tronothane, Tronothane hydrochloride
IUPAC Name 4-[3-(4-butoxyphenoxy)propyl]morpholine;hydrochloride
SMILES Cl.CCCCOc1ccc(OCCCN2CCOCC2)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C17H27NO3.ClH/c1-2-3-12-20-16-5-7-17(8-6-16)21-13-4-9-18-10-14-19-15-11-18;/h5-8H,2-4,9-15H2,1H3;1H
Formula Weight 329.1758
Chemical Name or Material Pramoxine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.