missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAP1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99413AF405
This item is not returnable.
View return policy
Description
PRAP1 Polyclonal antibody specifically detects PRAP1 in Human samples. It is validated for Western Blot
Specifications
| PRAP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 118471 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 405 | |
| PRO1195, proline-rich acidic protein 1, RP11-122K13.6, UPAMGC126792, Uterine-specific proline-rich acidic protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PRAP1. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction