missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAS40 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00 - € 560.70
Specifications
| Antigen | PRAS40 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18274363
|
Novus Biologicals
NBP2-54956 |
100 μL |
€ 593.00 € 560.70 / 100µL Save € 32.30 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659196
|
Novus Biologicals
NBP2-54956-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRAS40 Polyclonal specifically detects PRAS40 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| PRAS40 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Autophagy, Cancer, Growth and Development, Lipid and Metabolism, mTOR Pathway, Neuronal Cell Markers, Neuroscience, Tumor Suppressors | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 84335 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KSLPVSVPVWGFKEKRTEARSSDEENGPPSSPDLDRIAASMRALVLREAEDTQVFGDLPRPRLNTSDFQKLKRK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 40 kDa, AKT1 substrate 1 (proline-rich), MGC2865, proline-rich AKT1 substrate 1,40 kDa proline-rich AKT substrate | |
| AKT1S1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title