missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRB4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05584
This item is not returnable.
View return policy
Description
PRB4 Polyclonal antibody specifically detects PRB4 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| PRB4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| basic salivary proline-rich protein 4, Po, proline-rich protein BstNI subfamily 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 178-247 of human PRB4 (NP_002714.2). SPPGKPQGPPQQEGNKPQGPPPPGKPQGPPPAGGNPQQPQAPPAGKPQGPPPPPQGGRPPRPAQGQQPPQ | |
| 100 μg | |
| Cell Biology, GPCR, Signal Transduction | |
| 5545 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction