missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRC1 Polyclonal Antibody

PRC1 Polyclonal Antibody
Brand: Invitrogen PA534378
This item is not returnable.
View return policy
Description
PRC1 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
| PRC1 | |
| Polyclonal | |
| Unconjugated | |
| PRC1 | |
| anaphase spindle elongation 1 homolog; ASE1; carboxypeptidase C PRC1; Carboxypeptidase Y; Carboxypeptidase YSCY; CPD-Y; CPY1; D7Ertd348e; LBC1; MGC1671; MGC3669; prc1; protein regulating cytokinesis 1; protein regulator of cytokinesis 1; YMR297W | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 233406, 308761, 9055 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O43663, Q99K43 | |
| PRC1 | |
| A 19 amino acid synthetic peptide near the center of human PRC1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction