missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRCC Monoclonal Antibody (OTI5H7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812966S
This item is not returnable.
View return policy
Description
PRCC Monoclonal antibody specifically detects PRCC in Human, Mouse samples. It is validated for Western BlotSpecifications
| PRCC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92733 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5546, 94315 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5H7 | |
| Unconjugated | |
| Prcc | |
| LOW QUALITY PROTEIN: proline-rich protein PRCC; papillary renal cell carcinoma (translocation-associated); papillary renal cell carcinoma translocation-associated gene product; papillary renal cell carcinoma translocation-associated gene protein; PRCC; PRCC, proline rich mitotic checkpoint control factor; proline rich mitotic checkpoint control factor; proline-rich protein PRCC; RCCP1; si:ch211-210h11.8; TPRC; Unknown (protein for MGC:140094) | |
| Human recombinant protein fragment corresponding to amino acids 135-491 of human PRCC produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction