missing translation for 'onlineSavingsMsg'
Learn More

PRDM12 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18699931 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18699931 0.02 mL 0.02mL
18627612 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18699931 Supplier Novus Biologicals Supplier No. NBP2942520.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRDM12 Polyclonal antibody specifically detects PRDM12 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias PFM9, PR domain containing 12, PR domain zinc finger protein 12, PR domain-containing protein 12, PR-domain containing protein 12, PR-domain zinc finger protein 12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-330 of human PRDM12 (NP_067632.2). LGIFSKTWIKAGTEMGPFTGRVIAPEHVDICKNNNLMWEVFNEDGTVRYFIDASQEDHRSWMTYIKCARNEQEQNLEVVQIGTSIFYKAIEMIPPDQELLVWYGNSHNTFLGIPGVPGLEEDQKKNKHEDFHPADSAAGPAGRMRCVICHRGFNSRSNLRSHMRIHTLDKPFVCRFCNRRFSQSSTLRNHVRLHTGERPYKCQVCQSAYSQLAGLRAHQKSARHRPPSTAL
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 59335
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.