missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM14 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98845H
This item is not returnable.
View return policy
Description
PRDM14 Polyclonal antibody specifically detects PRDM14 in Human samples. It is validated for Western Blot
Specifications
| PRDM14 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 63978 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| MGC59730, PFM11, PR domain containing 14, PR domain zinc finger protein 14, PR domain-containing protein 14, PR-domain zinc finger protein 14 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PRDM14. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction