Learn More
PRDM14 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98845PE
Product Code. 18676245
  • € 407.00 / 0.10mL
Description
PRDM14 Polyclonal antibody specifically detects PRDM14 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rabbit
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PRDM14.
Antigen and protein A Affinity-purified
0.1 mL