missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRDM14 Monoclonal Antibody (647002)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA524292

Product Code. 15553982

  • € 476.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

In direct ELISAs, no cross-reactivity with recombinant mouse PRDM14, recombinant human (rh) ZNF24, rhZNF143, rhZNF206 or rhZNF281 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

PRDM14 encodes a member of the PRDI-BF1 and RIZ homology domain containing (PRDM) family of transcriptional regulators. The encoded protein may possess histone methyltransferase activity and plays a critical role in cell pluripotency by suppressing the expression of differentiation marker genes. Expression of this gene may play a role in breast cancer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRDM14
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q9GZV8
Prdm14
E. coli-derived recombinant human PRDM14 Ala2-Asp201.
100 μg
Primary
Human
Antibody
IgG1
Western Blot, Immunocytochemistry
647002
Unconjugated
Prdm14
LOC100365184; PFM11; PR domain 14; PR domain containing 14; PR domain containing 14-like; PR domain zinc finger protein 14; PR domain-containing 14; PR domain-containing protein 14; PR/SET domain 14; PRDM14
Mouse
Protein A/G
RUO
63978
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.