missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM16/MEL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71992
This item is not returnable.
View return policy
Description
PRDM16/MEL1 Polyclonal specifically detects PRDM16/MEL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PRDM16/MEL1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| PRDM16 | |
| A 17 amino acid synthetic peptide near the C-terminus of human PRDM16/MEL1. The immunogen is located within amino acids 1120 - 1170 of PRDM16/MEL1. | |
| 0.05 mg | |
| Chromatin Research | |
| 63976 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| MDS1/EVI1-like gene 1, MEL1KIAA1675PFM13MGC166915, PR domain containing 16, PR domain zinc finger protein 16, PR domain-containing protein 16, Transcription factor MEL1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human PRDM16 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction