missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRDM4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518312
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
The protein encoded by this gene is a transcription factor of the PR-domain protein family. It contains a PR-domain and multiple zinc finger motifs. Transcription factors of the PR-domain family are known to be involved in cell differentiation and tumorigenesis. An elevated expression level of this gene has been observed in PC12 cells treated with nerve growth factor, beta polypeptide (NGF). This gene is located in a chromosomal region that is thought to contain tumor suppressor genes.
Specifications
| PRDM4 | |
| Polyclonal | |
| Unconjugated | |
| PRDM4 | |
| 1700031E19Rik; 2810470D21Rik; AW552272; MGC45046; PFM1; PR domain 4; PR domain containing 4; PR domain zinc finger protein 4; PR domain-containing protein 4; PRDM4; PR-domain zinc-finger protein PFM1; SC-1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 11108, 72843 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q80V63, Q9UKN5 | |
| PRDM4 | |
| Synthetic peptide sequence (CSAVYSADESLSAHK) corresponding to the C-terminus amino acids of PRDM4 (aa 788-801). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction