missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRDM5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA596653

Product Code. 16539491

  • € 516.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MLGMYVPDRF SLKSSRVQDG MGLYTARRVR KGEKFGPFAG EKRMPEDLDE NMDYRLMWEV RGSKGEVLYI LDATNPRHSN WLRFVHEAPS QEQKNLAAIQ; Positive Samples: Mouse kidney, Mouse heart, Mouse lung, Mouse ovary, Rat kidney, Rat liver; Cellular Location: Nucleus.

PRDM5, a novel member of PR domain protein family, contains a PR-domain at the -NH2 terminus followed by 16 zinc finger motifs. It is found to have a role in cell differentiation and tumorigenesis. Reports suggest PRDM5 causes G2/M arrest and apoptosis upon infection of tumor cells and has been found to be silenced in breast, ovarian and lung cancers, suggesting a potential clinical significance.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRDM5
Polyclonal
Unconjugated
PRDM5
4432417F03Rik; 6530401I24Rik; AI197291; BCS2; E130112L17Rik; PFM2; PR domain 5; PR domain containing 5; PR domain zinc finger protein 5; PR domain-containing protein 5; PR/SET domain 5; PRDM5; unm hi61; un-named hi61
Rabbit
Affinity Chromatography
RUO
11107, 689788, 70779
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9CXE0, Q9NQX1
PRDM5
Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human PRDM5 (NP_0611692).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.