missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRDM9 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA538001
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
The PR domain is a protein-protein interaction module of about 100 amino acids. PR domain-containing proteins, such as PRDM9, are often involved in transcriptional regulation.
Specifications
| PRDM9 | |
| Polyclonal | |
| Unconjugated | |
| PRDM9 | |
| Histone-lysine N-methyltransferase PRDM9; MEISETZ; minisatellite binding protein 3 (115kD); MSBP3; PFM6; PR domain 9; PR domain containing 7; PR domain containing 9; PR domain zinc finger protein 9; PR domain-containing protein 9; Prdm7; Prdm9; Protein-lysine N-methyltransferase PRDM9; ZNF899 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 365155 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P0C6Y7 | |
| PRDM9 | |
| Peptide with sequence C-QEHFDSQNKNDK, from the internal region of rat Prdm9 (aa440-451). | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction