missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55087-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to PRDX3.
Prostaglandin-endoperoxide synthase (PTGS), also known as cyclooxygenase, is the key enzyme in prostaglandin biosynthesis, and acts both as a dioxygenase and as a peroxidase. There are two isozymes of PTGS: a constitutive PTGS1 and an inducible PTGS2, which differ in their regulation of expression and tissue distribution. This gene encodes the inducible isozyme. It is regulated by specific stimulatory events, suggesting that it is responsible for the prostanoid biosynthesis involved in inflammation and mitogenesis.Specifications
| PRDX3 | |
| Polyclonal | |
| Unconjugated | |
| PRDX3 | |
| Antioxidant protein 1, AOP 1, AOP1, HBC189, MER5, Peroxiredoxin 3, Peroxiredoxin III, PRDX3, PRO1748, Protein MER5 homolog, Prx III, PRX3, SP 22 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10935, 11757, 64371 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P20108, P30048, Q9Z0V6 | |
| Prdx3 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction