missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDX5 Monoclonal Antibody (OTI4B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811481S
This item is not returnable.
View return policy
Description
PRDX5 Monoclonal antibody specifically detects PRDX5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PRDX5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P30044 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 25824 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B12 | |
| Unconjugated | |
| PRDX5 | |
| ACR1; Alu corepressor 1; Alu co-repressor 1; Antioxidant enzyme B166; AOEB166; AOPP; B166; epididymis secretory protein Li 55; HEL-S-55; Liver tissue 2D-page spot 2D-0014IV; liver tissue 2D-page spot 71B; peroxiredoxin 5; peroxiredoxin 6; peroxiredoxin V; Peroxiredoxin-5, mitochondrial; peroxisomal antioxidant enzyme; peroxisomal membrane protein 20; PLP; Pmp20; PRDX5; Prdx6; Prx5; PrxV; prx-V; SBBI10; Thioredoxin peroxidase PMP20; thioredoxin reductase; TPx type VI | |
| Full length human recombinant protein of human PRDX5 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction