Learn More
PRDX6 Monoclonal Antibody (OTI11B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
€ 242.00 - € 540.87
Specifications
| Antigen | PRDX6 |
|---|---|
| Clone | OTI11B8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17508374
|
Origene Technologies
TA813658S |
30 μL |
€ 242.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17588364
|
Origene Technologies
TA813658 |
100 μL |
€ 540.87
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
PRDX6 Monoclonal antibody specifically detects PRDX6 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PRDX6 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| 1-Cys; 1-Cys peroxiredoxin; 1-Cys PRX; 1-cysPrx; 24 kDa protein; 9430088D19Rik; AA690119; Acidic calcium-independent phospholipase A2; aiPLA2; Antioxidant protein 2; Aop2; Brp-12; CP-3; epididymis secretory sperm binding protein Li 128m; GPx; HEL-S-128m; KIAA0106; Liver 2D page spot 40; Ltw4; Ltw-4; Lvtw-4; MGC46173; MSP23; NKEFA; Non-selenium glutathione peroxidase; NSGPx; ORF06; p29; PAG; PAGA; PAGB; peroxiredoxin 6; peroxiredoxin-6; Prdx5; Prdx6; PRX; Prx6; Red blood cells page spot 12; thiol-specific antioxidant protein; Tsa | |
| Full length human recombinant protein of human Prdx6 (NP_004896) produced in E.coli. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| PRDX6 |
| OTI11B8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| O08709, O35244, P30041 | |
| 11758, 94167, 9588 | |
| IgG1 | |
| Protein A/G | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.